missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ Hopen™ Polypropylene Coveralls

Product Code. 17700109 Shop All Medicom Products
Change view
Click to view available options
Color:
Blue
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Size Unit Size
17700109 Blue Large Pack of 25
17710109 Blue X-Large Pack of 25
17720109 Blue 2X-Large Pack of 25
17730109 Blue 3X-Large Pack of 25
17790099 Blue Medium Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17700109 Supplier Medicom™ Supplier No. 40.201BM/03
  • ONLINE EXCLUSIVE PRICE
    66.00 €
    43.45 € / Pack of 25
    Save 22.55 €
    34% Off
View all promotions
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Medicom™ Hopen™ Polypropylene Coveralls available in collared, or hooded styles

  • Hygiene Classic Polypropylene Coveralls protect the working environment from particles coming from the body of the operator
  • Protects the operator from the risks of soiling related to their purpose for use
  • Polypropylene fabric is light, and breathable
  • Elasticated at cuffs, waist, and ankles
  • Individually packaged for controlled hygiene and distribution
  • Category 1 Personal Protective Equipment

Specifications

Product Type Cleanroom Coveralls
Size Large
Gender Unisex
Material Polypropylene
Color Blue
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Elastic
Closure Type Zipper
Seam Type Stitched
Sterility Non-sterile
Certifications/Compliance Manufactured under an ISO 9001 Certified Quality System
Latex Free No
No. of Pockets None
For Use With (Application) Controlled Environments
Weight (Metric) 40 g/m2
Quantity 25/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.