missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ Hopen™ Simple White Coveralls

Product Code. 16544041 Shop All Medicom Products
Change view
Click to view available options
Neck Style:
Collared
Hooded
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Thickness:
40 g/m2
Unit Size:
Pack of 25
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Neck Style Size Thickness Unit Size
16544041 Collared Large 40 g/m2 Pack of 25
16534041 Collared Medium 40 g/m2 Pack of 25
16554041 Collared X-Large 40 g/m2 Pack of 25
16564041 Collared 2X-Large 40 g/m2 Pack of 25
16574041 Collared 3X-Large 40 g/m2 Pack of 25
16564011 Hooded Large 40 g/m2 Pack of 25
16574011 Hooded X-Large 40 g/m2 Pack of 25
16584011 Hooded 2X-Large 40 g/m2 Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 16544041 Supplier Medicom™ Supplier No. 40.101/03
  • ONLINE EXCLUSIVE PRICE
    56.00 €
    40.38 € / Pack of 25
    Save 15.62 €
    27% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Medicom™ Hopen™ Simple White Coveralls available in a variety of sizes, and styles to fit every need

  • Single use, white coveralls can be used for a wide variety of industrial needs
  • Individually packaged for hygiene, and controlled distribution purposes
  • Suitable to HACCP procedures

Specifications

Product Type Cleanroom Coveralls
Size Large
Gender Unisex
Material Polypropylene
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Closure Type Zipper
Seam Type Stitched
Sterility Non-sterile
Certifications/Compliance Manufactured under an ISO 9001 Certified Quality System
No. of Pockets None
Thickness 40 g/m2
For Use With (Application) Controlled Environments
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.