missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ Hopen™ Single Elastic Bouffant Cap

Product Code. 16574111 Shop All Medicom Products
Change view
Click to view available options
Color:
Blue
White
Unit Size:
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
16574111 White Pack of 1000
16584111 Blue Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16574111 Supplier Medicom™ Supplier No. 70.101
  • ONLINE EXCLUSIVE PRICE
    32.00 €
    23.53 € / Pack of 1000
    Save 8.47 €
    26% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Medicom™ Hopen™ Single Elastic Bouffant Cap provides full-head coverage to ensure user retention

  • Simple elastic model provides the user with good support by covering the entirety of their hair
  • Possibility of identification by color code
  • Intended to be worn by patients and caregivers in the operating room
  • Can be used as an object for controlling the environment in HACCP and various other industries

Specifications

Material Polypropylene
Color White
Disposable Single-use
Sterility Non-sterile
For Use With (Equipment) Operating Room PPE
Certifications/Compliance Manufactured under an ISO 9001 Certified Quality System
Packaging Type Bag
Dimensions (L x W) 44 x 50 cm
Weight (Metric) 12 g/m2
Construction Elasticated with Welded Ends
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.