missing translation for 'onlineSavingsMsg'
Learn More

Isofield™ Horizon Cleanroom Hood - ISO Class 4/5 Compatible

Product Code. 32016122
Change view
Click to view available options
Size:
Universal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size
32016122 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32016122 Supplier Isofield™ Supplier No. 80920
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Features an innovative postbox design with half-face elastic and a flat-top shape for excellent forehead coverage.

  • Non-sterile cleanroom hood
  • Innovative postbox hood design
  • Half-face elastic for additional comfort
  • Flat-top design ensures excellent forehead coverage
  • Aseptically folded
  • Class 1 for cleanliness
  • Static-dissipative
  • Tested against chemotherapy
  • Microporous polypropylene/polyethylene
  • 60 gsm, low-linting robust fabric

Technical and Documentation Compliance:

  • COA, COC available for download online
  • QR code on cartons allows for immediate document retrieval
  • Type PB[6]
  • Resistance to penetration by spray EN 13034:2015 + A1:2009 / ISO 17491-4:2008
  • Inward leakage of aerosols and fine particles EN ISO 13982-2:2004 / EN ISO 13982-1:2004
  • Permeation of liquids and gases ASTM F739-20
  • Particulate cleanliness - Helmke Drum Test: Class 1
  • Surface resistivity ASTM D257

Specifications

Product Type Cleanroom Hood
Size Universal
Material Polyethylene, Non-woven Polypropylene Laminate
Color White
Closure Type Ties
Disposable Single-use
Design Type Postbox
Latex Free Yes
Model Horizon
Seam Type Bound
Sterility Non-sterile
Thickness 2.6 mil
Certifications/Compliance (Europe) PPE Regulation (EU) 2016/425
Packaging Type Double-Bagged
Quantity 140 Pk.
Includes Horizon Cleanroom Hood
Construction Postbox Hood with Shoulder Coverage
For Use With (Application) GMP Cleanroom, Pharmaceutical, Biotechnology, Medical Device, Laboratory, and Electronics Manufacturing Environments
Hood Type Standard
Cleanroom Class Class 100 (ISO 5)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.