missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Horse anti-Goat IgG (H+L) ReadyProbes™ Secondary Antibody, Biotin

Product Code. 30245432
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30245432 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30245432 Supplier Invitrogen™ Supplier No. SA510421
  • ONLINE EXCLUSIVE PRICE
    267.00 €
    237.63 € / 50mL
    Save 29.37 €
    11% Off
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Horse Polyclonal Secondary Antibody

Recognizes both heavy and light chains (H+L). Suitable for use with goat, sheep, and bovine IgG primary antibodies No pipetting or dilutions required. Just tip and drip enough reagent to adequately cover the tissue or cells (typically 2-3 drops).

A Horse anti-Goat IgG antibody is an immunoglobulin produced by immunizing a horse with goat IgG. This generates a specific immune response in the horse, leading to the production of antibodies that can recognize and bind to goat IgG. In scientific assays, such as Western blotting, ELISA, or immunohistochemistry, this antibody is used as a secondary antibody to detect a goat-derived primary antibody bound to its target antigen. Typically, the horse anti-goat IgG antibody is conjugated to a reporter molecule (e.g., an enzyme like horseradish peroxidase or a fluorescent dye), which allows for the indirect detection and quantification of the antigen-antibody complex. This system amplifies the signal, making it easier to detect low-abundance targets.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat IgG (H+L)
Applications Immunohistochemistry, Immunocytochemistry, ELISA, In Situ Hybridization (ISH), Western Blot
Classification Polyclonal
Concentration 10 μg/mL
Conjugate Biotin
Formulation PBS with carrier protein and 0.08% sodium azide
Host Species Equine
Purification Method Affinity chromatography
Quantity 50 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Goat
Content And Storage 4°C
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.