missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Horse anti-Mouse IgG (H+L) Secondary Antibody, Biotin

Product Code. 11513350
Change view
Click to view available options
Quantity:
1.5 mg
Unit Size:
1.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11513350 1.5 mg 1.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11513350 Supplier Invitrogen™ Supplier No. 31806
  • ONLINE EXCLUSIVE PRICE
    229.00 €
    203.81 € / 1.5mg
    Save 25.19 €
    11% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Horse Polyclonal Secondary Antibody

Product # 31806 has been successfully used in Western blot, IF, ICC, IHC and ISH applications. Product # 31806 reacts with the heavy chains of mouse IgG and with the light chains common to most mouse immunoglobulins. This antibody cross-reacts with mouse IgM (50%) hamster IgG, rat IgG (12%) rat IgM (6%) dog IgG, guinea pig IgG, swine IgG (3%) Rhesus monkey IgG (1%). It has minimal cross-reactivity with immunoglobulins from other species.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgG (H+L)
Applications ELISA, Immunohistochemistry, In Situ Hybridization (ISH), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate Biotin
Formulation PBS with 3mg/mL BSA and 0.08% sodium azide; pH 7.8
Host Species Equine
Purification Method Affinity chromatography
Quantity 1.5 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability.
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.