missing translation for 'onlineSavingsMsg'
Learn More

HUBER Hose for Cooling Water

Product Code. 10769253
Click to view available options
For Use With (Equipment):
Cooling Water PZ-90-1-G1
Cooling Water PZ-90-1-G1/2
Cooling Water PZ-90-1-G3/4
Cooling Water PZ-90-1.5-G1
Cooling Water PZ-90-1.5-G1/2
Cooling Water PZ-90-1.5-G3/4
Cooling Water PZ-90-2-G1
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10769253

Brand: HUBER 16858

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Braided hose

  • Braided hose
  • Min. temp.: -20°C
  • Max. temp.: 90°C

Specifications

For Use With (Equipment) Cooling Water PZ-90-1.5-G1
Temperature Range -20°C to +90°C
Type Hose
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.