missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HotStart-IT Binding Protein

Product Code. 15643239
Click to view available options
Quantity:
400 μg (200 x 25 μL rxns)
Unit Size:
400µg
This item is not returnable. View return policy

Product Code. 15643239

Brand: Thermo Scientific™ 71194400UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HotStart-IT Binding Protein is the active component in a novel hot start technology called primer sequestration.

HotStart-IT Binding Protein is the active component in the novel hot start technology known as primer sequestration. In primer sequestration, hot start PCR methods reduce or eliminate non-specific primer-extension products formed at lower temperatures during assembly of PCR reactions. At these less stringent annealing temperatures, primers may bind non-specifically, which often leads to unwanted amplification products and primer-dimers. To resolve this problem, we have produced a high-quality DNA binding protein that is especially useful at sequestering primers at lower temperatures, making them unavailable for use by a polymerase (see figure).

Features

  • Room temperature reaction setup
  • Minimizes amplification of non-specific products and primer-dimers (see figure)
  • Ideal for complex templates and multiplex reactions
  • No extensive heating step required
  • Free from detectable non-specific nucleases
  • Technology is portable to a polymerase of choice

This primer-sequestration technique effectively blocks DNA synthesis from mis-priming events at lower temperatures. Following the initial denaturation step during PCR, the protein is inactivated, and the primers are free to participate in the amplification reaction.

HotStart-IT Binding Protein performs well in many standard PCR reaction buffers and has been designed for PCR applications that demand high specificity and sensitivity. PCR with Taq DNA Polymerase and HotStart-IT Binding Protein shifts production of primer-dimers to a specific target of 306 bp from 1 ng of human genomic DNA relative to Taq DNA Polymerase alone.

Application

  • Hot-Start PCR amplification

Notes

  • One microgram of HotStart-IT Binding Protein sequesters about 5 pmol of primers.
  • HotStart-IT Binding Protein is sourced from recombinant protein expressed in E. coli.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Hot-start PCR
Content And Storage • HotStart-IT Binding Protein, 400 μg

Store at -20°C.
Shipping Condition Dry Ice
Compatible Buffer 1X PCR Reaction Buffer
Quantity 400 μg (200 x 25 μL rxns)
Product Type HotStart-IT Binding Protein

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.