missing translation for 'onlineSavingsMsg'
Learn More

Howard Leight™ Bilsom™ 303L Disposable Earplug Refill

Product Code. 11847030
Change view
Click to view available options
Quantity:
200/Pk.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11847030 200/Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11847030 Supplier Honeywell Safety Products Supplier No. 1006186
  • ONLINE EXCLUSIVE PRICE
    65.73 €
    44.18 € / Pack of 1
    Save 21.55 €
    32% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Wears more comfortably than other foam earplugs over long periods of time. Sperian™ Howard Leight™ Bilsom™ 303L Disposable Earplug Refills are ideal for medium to high-noise environments.

  • Polyurethane foam expands gently in ear canal, resulting in less expansion pressure
  • Smooth, soil-resistant skin prevents dirt build-up
  • Tapered bullet-shaped design is easier to roll-down and insert, adapts more evenly to ear canal
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Certifications/Compliance EN 352-2: 2002, ISO 9001/2000
Size Universal
For Use With (Application) For workers who need the suppression of loud noises
Quantity 200/Pk.
Plug Shape Bullet
Product Type Ear Plugs
Disposable Single-use
Material Polyurethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.