missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Howard Leight™ Leightning L2FHV Hi-Visibility Folding Ear Muff

Product Code. 12635173
Change view
Click to view available options
L2F; Folding; High-Vis Lime, Over-the-Head
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12635173 L2F; Folding; High-Vis Lime, Over-the-Head
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 12635173 Supplier Honeywell™ Supplier No. 1013942
Quantity Price
1 25.34 € / Pack of 1
20 22.70 € / Pack of 1
In Stock
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Total visibility, performance and durability

  • Bright green earcups provide high visibility and contrast
  • Reflective padded headband illuminates under light for increased visibility and safety
  • Ideal to wear at night, during the day, in low light and under inclement weather conditions
  • Patented Air Flow Control™ technology provides optimal attenuation across all frequencies
  • Robust steel headband withstands demanding use, especially in tough environments
  • Snap-in ear cushions make replacement quick and easy
TRUSTED_SUSTAINABILITY

Specifications

Color High-visibility Lime
ANSI S3.1974 Yes
CSA Z94.2 Class BL
Cushion Color Black
Headband Material Foam
For Use With (Application) Construction, Manufacturing, Mining, Municipal Services, Oil and Gas, Pharmaceutical, Transportation
High Visibility Yes
Model L2FHV
Noise Reduction Rating (NRR) 27 dB
Product Type Ear Muffs
Disposable Reusable
Headband Color Gray
Dielectric No
Style Over-the-Head
Foldable Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.