Learn More
Honeywell™ Howard Leight Max (Uncorded) Earplugs
Product Code. 12666123
Click to view available options
:
Orange, SNR: 37 dB
Product Code. 12666123
Brand: Honeywell™ 3301161
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Soft foam earplugs for superior, long-wearing comfort and solid performance.
  • Soft foam delivers superior long-wearing comfort and solid performance
  • Bell-shaped design delivers maximum in-ear comfort without backing out of the ear canal, easier to insert
  • Smooth, soil-resistant exterior prevents dirt or other substances from accumulating on skin or entering ear canal
  • Superior noise-blocking performance in a single-use earplug
  • Enhanced contoured shape is easier to insert and resists backing out of the ear canal, while its softer foam delivers superior long-wearing comfort - delivering solid performance that your workers can depend on
TRUSTED_SUSTAINABILITY
Specifications
Color Orange
Certifications/Compliance (Europe) EC Category PPE: 2, QA: ISO 9001/2000, EC Certificate: 1219, EC Attestation: 040366
Noise Reduction Rating (NRR) SNR: 37 dB
Plug Shape Bell
Disposable Single-use
Material Polyurethane