missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Thunder T3 Ear Muff Hygiene Kit

Product Code. 12615023
Change view
Click to view available options
Includes:
2 Replacement ear cushions, 2 Foam inserts
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
12615023 2 Replacement ear cushions, 2 Foam inserts Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12615023 Supplier Honeywell™ Supplier No. 1010976
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extends performance and lifetime of the Thunder T3 Ear Muff. Includes one pair of snap-in ear cushions and one pair of foam ear cup inserts.

  • Hygiene Kit to be used with Thunder T3 Ear Muff only
  • Extends performance and lifetime of Ear Muff
  • Recommended replacement every 6 months with general use
  • Snap-in cushions for quick and easy replacement
  • Includes one pair of snap-in ear cushions and one pair of foam ear cup inserts
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Howard Leight™ Ear Muffs
For Use With (Application) For high attenuating noise blocking ear muffs
Product Type Thunder Ear Muff Hygiene Kit
Style Snap-in ear
Includes 2 Replacement ear cushions, 2 Foam inserts
compliance-icons
Product Identifier
  • Immersion oil
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H315-Causes skin irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Hexanedioic acid, bis[(octahydro-4,7-methano1H-inden-5-yl)methyl] ester,benzyl benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.