missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAT™ HPLC Supply and Waste Set, V2.0, Suitable for All HPLC Systems
Shop All SCAT Europe Products
missing translation for 'orderingAttributeHoverText'
Includes:
4 x Safety Cap III, GL45, 4 x air valve, 1 x Safety Waste Cap, S60/61, electrostatic conductive, 1 x Exhaust filter M, V3.0, with splash guard and change indicator, 1 x 5 liter canister, PE-HD-EL, 1 x Grounding cable, 2 x clamp, 1.5 m length, 4 x Laboratory bottle DURAN, GL45, 1000 mL, clear glass and round, with cap and pouring ring, 5 x PFA-fitting with integrated ferrule, 2.3 mm OD, violet, 5 x PFA fitting with integrated ferrule, 1.6 mm OD, green, 10 x PFA fitting with integrated ferrule, 3.2 mm OD, blue, 10 x PFA blind plug, 2 x connector, straight, 6 to 8 mm ID, 1 x connector, curved, 5 to 11.5 mm ID, 1 x connector, angled, 9.5 to 10 mm ID
4 x Safety Cap III, GL45, 8 x air valve, 1 x Safety Waste Cap, GL45, 1 x Exhaust filter L, V3.0, with splash guard and change indicator, service life 12 months, 1 x 5 litre canister, PE-HD, 4 x Laboratory bottle DURAN, GL45, 1000 ml, clear glass and round, with cap and pouring ring , 5 x PFA-fitting with integrated ferrule, 2.3 mm OD, violet, 5 x PFA fitting with integrated ferrule, 1.6 mm OD, green, 10 x PFA fitting with integrated ferrule, 3.2 mm OD, blue, 10 x PFA blind plug, 2 x connector, straight, 6 - 8 mm ID, 1 x connector, curved, 5.0 - 11.5mm ID, 1 x connector, angled, 9.5 - 10 mm ID
4 x Safety Cap III, GL45, 8 x air valve, 1 x Safety Waste Cap, S60/61, electrostatic conductive, 1 x Exhaust filter L, V3.0, with splash guard and change indicator, service life 12 month, 1 x 5 litre canister, PE-HD-EL, 1 x Grounding cable, 2x clamp, L = 1,5 m, 4 x Laboratory bottle DURAN, GL45, 1000 ml, clear glass and round, with cap and pouring ring , 5 x PFA-fitting with integrated ferrule, 2.3 mm OD, violet, 5 x PFA fitting with integrated ferrule, 1.6 mm OD, green, 10 x PFA fitting with integrated ferrule, 3.2 mm OD, blue, 10 x PFA blind plug, 2 x connector, straight, 6 - 8 mm ID, 1 x connector, curved, 5.0 - 11.5mm ID, 1 x connector, angled, 9.5 - 10 mm ID
4 x Safety Cap III, V2.0, GL45, 1 x Safety Waste Cap, V2.0, GL45, 1 x Exhaust filter M, V2.0, 4 x Laboratory bottle DURAN, GL45, 1000 mL, 4 x air valve, 5 x PFA fitting with integrated ferrule, 2.3 mm, violet, 5 x PFA fitting with integrated ferrule, 1.6 mm, green, 10 x PFA fitting with integrated ferrule, 3.2 mm, blue, 10 x Blanking plugs, 1.6/2.3/3.2 mm, colorless, 2 x Tube connector, straight 6 - 8 mm, 1 x Tube connector, curved, 5.0 - 11.5 mm, 1 x tube connector, angled, 9.5 - 10 mm
missing translation for 'unitSize'
1 set
1 sheet
Each
Descrizione
- Protects your system from contamination
- Keeps solvent mixtures stable
- High performance materials with excellent chemical resistance
The safety set connects tubing safely and blocks hazardous vapours. Air valves and exhaust filters provide safe pressure equalisation without under- or overpressure in solvent bottles and waste containers
Specifica
Specifica
| Description | Non-Electrostatic Conductive |
| For Use With (Equipment) | Any HPLC System |
| Type | HPLC System |
| Solvent Channels | 8 |
| Includes | 4 x Safety Cap III, V2.0, GL45, 1 x Safety Waste Cap, V2.0, GL45, 1 x Exhaust filter M, V2.0, 4 x Laboratory bottle DURAN, GL45, 1000 mL, 4 x air valve, 5 x PFA fitting with integrated ferrule, 2.3 mm, violet, 5 x PFA fitting with integrated ferrule, 1.6 mm, green, 10 x PFA fitting with integrated ferrule, 3.2 mm, blue, 10 x Blanking plugs, 1.6/2.3/3.2 mm, colorless, 2 x Tube connector, straight 6 - 8 mm, 1 x Tube connector, curved, 5.0 - 11.5 mm, 1 x tube connector, angled, 9.5 - 10 mm |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto