missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ HPLC Starter Kits I, Green Line

Product Code. 31454224 Shop All SCAT Europe Products
Change view
Click to view available options
Material:
PEEK
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material
31454224 PEEK
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31454224 Supplier SCAT™ Supplier No. 499200
  • ONLINE EXCLUSIVE PRICE
    383.00 €
    296.10 € / 1 sets
    Save 86.90 €
    22% Off
View all promotions
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Green Line Safety Caps stand for a conscious decision not to use PFAS, without compromising on quality and safety.

  • PFAS-free - future-proof and sustainable: The Green Line completely dispenses with PFAS and uses PEEK as a fluorine-free high-performance plastic that guarantees sustainability and future-proofing.
  • Full regulatory safety: Green Line products eliminate risks posed by upcoming PFAS bans and meet the highest environmental and ESG requirements.
  • Technical quality without compromise: The safety caps are 1:1 compatible with existing systems, chemically resistant, autoclavable and absolutely leak-proof in accordance with the SCAT standard.
  • Visibly sustainable: With their green coloring, the products make sustainability visible and demonstrate the laboratory's commitment to a PFAS-free future.
  • Development with foresight Green Line secures laboratory processes against regulatory risks in the long term and supports sustainable corporate strategies.
  • Product diversity and cost-effectiveness: The series offers a wide range of variants, starter kits and accessories for flexible, cost-effective and ready-to-use HPLC solutions.
  • In summary, SCAT Green Line stands for PFAS-free, regulatory safe, technically powerful and visibly sustainable laboratory solutions - without compromising on quality, tightness or compatibility. While conventional caps are based on PTFE, a representative of the so-called perpetual chemicals, the Green Line uses PEEK, a completely fluorine-free high-performance plastic with excellent chemical and thermal properties.

Specifications

For Use With (Application) PFAS-free analyses
For Use With (Equipment) GreenLine Portfolio
Type Safety Caps Kit
Material PEEK
Includes 3 x Safety Cap I, GL 45, PEEK, 1 x Safety Cap II, GL 45, PEEK, 4 x air valve, 4 x change indicator, 5 x PE-fitting 1.6 mm OD, 5 x PE-fitting 2.3 mm OD, 5 x PE-fitting 3.2 mm OD, 1 x PE-blind plug, 4 x labelling field
Quantity 4 Caps
Color Green
Thread Size GL45
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.