Learn More
STEMCELL Technologies hPSC Genetic Analysis Kit
Product Code. 15851657
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. Quantity Unit Size
15851657 1 Kit 1 set
1 options
Product Code. 15851657 Supplier STEMCELL Technologies Supplier No. 07550
 Restricted Item
Estimated Shipment: 25-11-2026
Lägg i varukorgen
Lägg i varukorgen
missing translation for 'validMainframeMsg'
Denna artikel kan inte returneras. Se returpolicy
qPCR analysis kit for detecting the majority of karyotypic abnormalities reported in human ES and iPS cells
  • Quickly and cost-effectively detect the 8 most common karyotypic abnormalities reported in human pluripotent stem cells (hPSCs)
  • This qPCR-based kit enables the genetic screening of multiple hPSC lines and contains enough material to analyze 20 individual samples in triplicate
  • It uses double-quenched probes with a 5-carboxyfluorescein (5-FAM) dye to give superior performance over other, single-quenched probes
  • The kit includes qPCR Master Mix, ROX Reference Dye, Buffer, and primer/probe mixes designed to detect the critical minimal regions of the 8 most commonly mutated regions, as well as a primer/probe mix to a control location in the genome
  • A genomic DNA control is also provided with demonstrated normal copy number over the regions of interest
Specifikationer
Description Pluripotent Stem Cells
Quantity 1 Kit
For Use With (Application) Real Time PCR (qPCR)
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.