missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ HRM Software Module for ViiA™ 7 System

Product Code. 15959222 Shop All Applied Biosystems Products
Change view
Click to view available options
One software CD
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15959222 One software CD
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15959222 Supplier Applied Biosystems™ Supplier No. 4453724
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Expands the functionality of the ViiA7 system, allowing High Resolution Melting (HRM) analysis to be performed on PCR amplicons.

Expands the functionality of the ViiA7 system, allowing High Resolution Melting (HRM) analysis to be performed on PCR amplicons. HRM analysis can discriminate genetic variation by amplicon length, GC content, sequence variability or strand complementarity.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage One Software CD
For Use With (Equipment) ViiA™ 7 System
Product Line ViiA
Display Type Touchscreen
Software Type Primer Design, Genotyping
Operating System Windows
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.