missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech HRP-conjugated Streptavidin
Shop All Proteintech Products
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
Specifications
Specifications
| Conjugate | HRP |
| Formulation | 0.01 M Sodium phosphate, 0.25 M NaCl, 50% glycerol, pH7.6 with 3 mg/ml BSA |
| Quantity | 500 μL |
| Content And Storage | Store at -20°C. Stable for one year after shipment. |
| Form | Liquid |
| For Use With (Application) | ELISA, Western Blot |
| Purity or Quality Grade | The protein Streptavidin is purified from Streptomyces avidinii. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction