missing translation for 'onlineSavingsMsg'
Learn More

Proteintech HRP -Protein A

Product Code. 17690297 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17690297 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17690297 Supplier Proteintech Supplier No. SA0000118500UL
 Restricted Item
Estimated Shipment: 20-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

HRP -Protein A

Specifications

Quantity 500 μL
Form Liquid
For Use With (Application) Western Blot
Content And Storage Store at -20°C. Stable for one year after shipment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.