missing translation for 'onlineSavingsMsg'
Learn More

BTX™ HT 96-well Electroporation Plate
GREENER_CHOICE

Product Code. 15487310
Change view
Click to view available options
Size (Metric) Gap:
2 mm
4 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size (Metric) Gap unitSize
15487310 4 mm Each
15477310 2 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487310 Supplier BTX™ Supplier No. 450452
Add to basket
Add to basket
This item is not returnable. View return policy

HT 96-well Electroporation Plate for use with ECM 630 and ECM 830 Electroporation Systems.

  • 96-well disposable electroporation plate
  • 2 mm gap
  • 100 μL
  • Coated
  • 1 plate

Specifications

Format 96-well
For Use With (Application) Transfection of Mammalian Cell, Transformation of Bacteria and Yeast, siRNA and cDNA Library Screening
For Use With (Equipment) ECM 630 HT, ECM 830 HT and Gemini X2 HT Electroporation Systems
Product Line BTX Electroporation
Product Type Electroporation Consumable
Description 96-well Disposable Electroporation Plates, 4 mm gap, 200 μl, 1 plate
Electroporation Chamber 96-well Electroporation Plate
Quantity 1 Ea.
Size (English) Gap 0.15 in.
Size (Metric) Gap 4 mm
Volume (Metric) 200 μL (Working)
Sterility Sterile
Connection Type Banana to Pin Tip
Pulse Form Exponential Decay Wave, Square Wave
High-throughput Compatibility Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.