missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ HT ExoSAP-IT™ High-Throughput PCR Product Cleanup
Click to view available options
No. of Reactions:
480 Reactions
5760 Reactions
Unit Size:
Each
Description
- One-tube PCR clean-up — Add HT ExoSAP-IT reagent directly to PCR product
- Exceptional accuracy — Achieve high quality data even with long read lengths
- 100% sample recovery — No loss of PCR products regardless of the fragment size
- Simple, single-step — Replaces the multiple steps and wasted time inherent with beads or columns
- High-throughput processing — Even faster time to results with a low viscosity formulation, allowing robotic pipetting
- Scalable - Treat reaction volumes from 5 μL to 5 L
- Convenient packaging — Available in 8-tube strips and 12 strips in a 96-well plate
- Highly stable — Retains full functional activity at 4°C for one week and at 25°C for 8 hours
Specifications
Specifications
| Content And Storage | Shipped on dry ice. Store at -20°C. |
| Description | PCR Product Clean-up, Single Step, High-throughput Processing, Scalable, Specifically Designed for the Unique Requirements of High-throughput, Automated Platforms and Multi-channel Pipettes |
| Format | 8 Tube Strip, 96-well Plate, Tube |
| For Use With (Application) | PCR Purification |
| No. of Reactions | 5760 Reactions |
| Product Type | PCR Product Clean-up |
| Quantity | 5,760 reactions |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction