missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ HTH 01-015
GREENER_CHOICE

Product Code. 18778328
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18778328

Brand: Tocris Bioscience™ 5622/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective NUAK1 inhibitor

HTH 01-015 is a potent and selective NUAK1 inhibitor (IC50 = 100 nM). Exhibits no significant inhibition against a panel of 139 kinases, including ten AMPK family members. Inhibits NUAK1-mediated MYPT1 phosphorylation. Also inhibits cell proliferation in U2OS cells in vitro, to a similar extent as NUAK1 knockdown models.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5,13-Dihydro-4,5,13-trimethyl-2-[[1-(4-piperidinyl)-1H-pyrazol-4-yl]amino]-6H-naphtho[2,3-e]pyrimido[5,4-b][1,4]diazepin-6-one
CAS 1613724-42-7
Quantity 10 mg
Target Other Kinase Inhibitors
Molecular Formula C26H28N8O
Formula Weight 468.55
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.