missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals HTLV Blot 2.4 (RUO)

Product Code. 30694935 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
18 Tests/Kit
36 Tests/Kit
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30694935 36 Tests/Kit 1 set
30694932 18 Tests/Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30694935 Supplier MP Biomedicals Supplier No. 11082036
  • ONLINE EXCLUSIVE PRICE
    1294.95 €
    1233.29 € / 1 sets
    Save 61.66 €
    4% Off
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The Blot is a qualitative enzyme immunoassay for the in vitro detection of antibodies to HTLV-1 and HTLV-2 in human serum or plasma.

  • The nitrocellulose strips are precoated with HTLV viral proteins derived from native inactivated viral particles and genetically engineered proteins.
  • Individual nitrocellulose strips are incubated with diluted serum or plasma specimens and controls.
  • Specific antibodies to HTLV-I/II, if present in the specimen will bind to the HTLV-I/II proteins on the strips.
  • The strips are washed to remove unbound materials while antibodies that bind specifically to the HTLV proteins can be visualized using a series of reactions with goat anti-human IgG conjugated with alkaline phosphatase and the substrate, BCIP/NBT.
  • Each strip also includes an internal sample addition control to minimize the risk of false negatives due to operational errors
  • The possible serological profiles defined by the test include the following: HTLV Seropositive, HTLV-I Seropositive, HTLV-II Seropositive, Seronegative or Indeterminate.
  • It is intended for use as a more specific supplemental test on human serum or plasma specimens found repeatedly reactive using screening procedures like ELISA.
  • Research use only.
  • For professional use only.
  • Human serum and plasma.

Specifications

Product Type Western Blot Kit
Detection Method Colorimetric
For Use With (Application) Immunoblot (Western)
Membrane Compatibility Nitrocellulose
Quantity 36 Tests/Kit
Substrate Alkaline Phosphatase
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.