missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human A2M native Protein from human blood

Product Code. 16262170
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16262170 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16262170 Supplier Abnova™ Supplier No. P4913.1mg

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for SDS-PAGE

Alpha-2-macroglobulin is a protease inhibitor and cytokine transporter. It inhibits many proteases, including trypsin, thrombin and collagenase. A2M is implicated in Alzheimer disease (AD) due to its ability to mediate the clearance and degradation of A-beta, the major component of beta-amyloid deposits. [provided by RefSeq]

Specifications

Concentration 2-4 mg/mL
For Use With (Application) SDS-PAGE
Formulation Lyophilized from 50mM phosphate buffer, 100mM NaCl, 500μg glycine, pH 7.4
Gene ID (Entrez) 2
Molecular Weight (g/mol) 725kDa
Name A2M (Human) Native Protein
Preparation Method Native protein purified from human blood
Purification Method Saltprecipitation/Chelate Chromatography/Gelfiltration
Quantity 1 mg
Storage Requirements Store at -80°C. After reconstitution with deionized water, store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CPAMD5/DKFZp779B086/FWP007/S863-7
Common Name A2M
Gene Symbol A2M
Species Human
Protein Tag None
Expression System Native protein purified from human blood
Form Lyophilized
Purity or Quality Grade >90% by SDS-PAGE
Show More Show Less
Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.