missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human ACE Quantikine ELISA Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
The Human ACE Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human ACE/CD143 in cell culture supernates (50 ul), serum (10 ul), heparin plasma (10 ul), saliva (25 ul).
Specifications
Specifications
| Assay Range | 0.78 to 50 ng/mL |
| Assay Sensitivity | 0.051 ng/mL |
| Conjugate | HRP |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Cell Culture Supernatant, Plasma (Heparin), Saliva, Serum |
| Specificity | Natural and Recombinant Human ACE. In addition to soluble ACE, this kit recognizes membrane-bound forms from lysates of cultured cells and supernates. |
| Sufficient For | 1 x 96-well Plate |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 1636 |
| Gene Symbol | ACE |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction