missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ADAM12 S Isoform Recombinant Protein

Product Code. 11574211
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. Quantity unitSize
11574211 1 mg 1mg
1 options
This item is not returnable. View return policy

Product Code. 11574211

Brand: Invitrogen™ RP75523

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This product contains purified recombinant human ADAM12 S Isoform protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number O43184
Concentration 0.27 mg/mL
For Use With (Application) Control, Western Blot
Formulation 25 mM sodium acetate with 50% glycerol and no preservative; pH 4.8
Gene ID (Entrez) 8038
Molecular Weight (g/mol) 62.16 kDa
Name Human ADAM12 S Isoform
pH Range 4.8
Quantity 1 mg
Source E. coli
Immunogen N/A
Regulatory Status RUO
Gene Alias a disintegrin and metalloproteinase; ADAM; ADAM 12; ADAM metallopeptidase domain 12; ADAM12; ADAM12-OT1; ADAMs; CAR10; Disintegrin and metalloproteinase domain-containing protein 12; MCMP; MCMPMltna; meltrin-alpha; metalloendopeptidases; metalloprotease-disintegrin 12 transmembrane; MLTN; MLTNA; UNQ346/PRO545
Common Name ADAM12 S Isoform
Gene Symbol ADAM12
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Protein Tag His-tag
Sequence Amino acids Glu208- Ile738 containing an N-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.