missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Adrenal Whole Tissue Lysate (Adult Whole Normal)

Product Code. 18745953
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18745953 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18745953 Supplier Novus Biologicals™ Supplier No. NB82059171
  • ONLINE EXCLUSIVE PRICE
    670.00 €
    633.15 € / 1mg
    Save 36.85 €
    5% Off
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The Human Adrenal Whole Tissue Lysate (Adult Whole Normal) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot
Tissue Endocrine
Quantity 1 mg
Storage Buffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Test Specificity Total Protein - Human Adult Normal Tissue: Adrenal
Cellular Fraction Whole
Tissue Condition Normal
Cross Reactivity Fas Receptor/TNFRSF6/CD95
Host Species Human
Concentration 5 mg/mL
Product Type Human Adrenal Whole Tissue Lysate (Adult Normal)
Content And Storage -80°C. Avoid freeze-thaw cycles.
Life Stage Adult
Protein State Native
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.