Learn More
R&D Systems™ Human ALDH1A3 (NP_000684.2) VersaClone cDNA
Product Code. 18701307
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. Quantity Unit Size
18701307 10 μg 10µg
1 options
Product Code. 18701307 Supplier R&D Systems™ Supplier No. RDC3658
  • ONLINE EXCLUSIVE PRICE
    287.00 €
    270.90 € / 10µg
    Save 16.10 €
    5% Off
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Gene ALDH1A3
Gene ID (Entrez) 220
Gene Alias aldehyde dehydrogenase 1 family, member A3, Aldehyde dehydrogenase 6, aldehyde dehydrogenase family 1 member A3, ALDH1A6, ALDH6acetaldehyde dehydrogenase 6, EC 1.2.1, EC 1.2.1.5, RalDH3, RALDH-3, Retinaldehyde dehydrogenase 3
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.