missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human alpha-Thrombin (DFP) Native Protein

Product Code. 11574131
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11574131 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11574131 Supplier Invitrogen™ Supplier No. RP43101
  • ONLINE EXCLUSIVE PRICE
    154.00 €
    137.06 € / 100µg
    Save 16.94 €
    11% Off
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Native Protein

RP-43101 contains native human a-Thrombin (DFP) protein. RP-43101 may be used as a positive control in many immunological assays. The product is of human blood/plasma origin. Although the starting material was tested prior to initiation of the manufacturing process, and was found negative or nonreactive for anti-HIV-1/2, HIV-1 antigen(s), HBsAg, STS, anti-HCV, anti-HBcore and anti-HTLV I & II, extreme caution should be used when handling this material as there is a margin of error in all tests. This product is to be used for IN VITRO research purposes only, and is not intended for clinical or diagnostic use!.

Note: Antibody lot-to-lot variability may occur and dilution optimization of each production lot in specific application is recommended.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P00734
Concentration 1.9 mg/mL
For Use With (Application) Neutralization
Formulation 50% water with no preservative
Gene ID (Entrez) 2147
Molecular Weight (g/mol) 36.7 kDa
Name Human alpha-Thrombin (DFP)
pH Range 7.4
Purification Method SDS-PAGE
Quantity 100 μg
Source Human
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Activation peptide fragment 1; Activation peptide fragment 2; bovine prothrombin protein; Coagulation factor II; coagulation factor II (thrombin); coagulation factor II [thrombin]; coagulation factor II, thrombin; F2; prepro-coagulation factor II; preprothrombin; prothrombin; prothrombin B-chain; PT; RPRGL2; serine protease; THPH1; thrombin; Thrombin heavy chain; Thrombin light chain
Common Name alpha-Thrombin
Gene Symbol F2
Product Type Protein
Conjugate Unconjugated
Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Form Liquid
Protein Subtype Native
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.