missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Angiostatin Native Protein

Product Code. 11514171
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11514171 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11514171 Supplier Invitrogen™ Supplier No. RP43140
  • ONLINE EXCLUSIVE PRICE
    900.00 €
    801.00 € / 500µg
    Save 99.00 €
    11% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Native Protein

RP-43140 contains native human Angiostatin protein. RP-43140 may be used as a positive control in many immunological assays. The product is of human blood/plasma origin. Although the starting material was tested prior to initiation of the manufacturing process, and was found negative or nonreactive for anti-HIV-1/2, HIV-1 antigen(s), HBsAg, STS, anti-HCV, anti-HBcore and anti-HTLV I & II, extreme caution should be used when handling this material as there is a margin of error in all tests. This product is to be used for IN VITRO research purposes only, and is not intended for clinical or diagnostic use!.

The protein encoded by this gene is a secreted blood zymogen that is activated by proteolysis and converted to plasmin and angiostatin. Plasmin dissolves fibrin in blood clots and is an important protease in many other cellular processes while angiostatin inhibits angiogenesis. Defects in this gene are likely a cause of thrombophilia and ligneous conjunctivitis. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P00747
Concentration 4.7 mg/mL
For Use With (Application) Neutralization
Formulation 0.02M HEPES with 0.15M NaCl and no preservative; pH 7.4
Gene ID (Entrez) 5340
Molecular Weight (g/mol) 50 kDa
Name Human Angiostatin
pH Range 7.4
Purification Method SDS-PAGE
Quantity 500 μg
Source Human
Storage Requirements -20°C
Regulatory Status RUO
Gene Alias Activation peptide; AI649309; Angiostatin; Microplasmin; Pg; plasmin; plasmin heavy chain A; Plasmin heavy chain A, short form; Plasmin light chain B; plasminogen; PLG
Common Name Angiostatin
Gene Symbol PLG
Product Type Protein
Conjugate Unconjugated
Species Human
Content And Storage -20°C
Form Liquid
Protein Subtype Native
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.