missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human ANGPTL-7 Recombinant Protein, PeproTech®

Code produit 17800693
Modifier l'affichage
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
Conditionnement:
100µg
1mg
20µg
250µg
500µg
5µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity Conditionnement
17800693 500 μg 500µg
17800583 100 μg 100µg
17839932 1 mg 1mg
17811483 20 μg 20µg
17820703 250 μg 250µg
17841273 5 μg 5µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 17800693 Fournisseur Gibco™ Code fournisseur 13022500UG
  • ONLINE EXCLUSIVE PRICE
    3205.00 €
    2307.60 € / 500µg
    Save 897.40 €
    28% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Ajouter au panier
Ajouter au panier

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

Recombinant Human ANGPTL-7 is a glycoprotein that migrates by SDS-PAGE analysis at an apparent molecular weight of 40-50 kDa under reduced conditions, and contains 328 amino acid residues including a C-terminal His-tag. The calculated molecular weight of Recombinant Human ANGPTL-7 is 38.2 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Angptl7 (angiopoietin-like 7), alternately known as cornea-derived transcript 6 protein (CDT6) or AngX, is a 346 amino acid secreted protein that is expressed specifically in the stromal layer of cornea. Angptl7 forms disulfide-linked homotetramers via coiled-coil interactions and contains one fibrinogen C-terminal domain. Overexpressed in patients with glaucoma, Angptl7 serves as a potential therapeutic target for glaucoma and has been found to increase collagen expression. The gene encoding Angptl7 maps to human chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome.
TRUSTED_SUSTAINABILITY

Spécification

Accession Number O43827
For Use With (Application) Binding Assay, Control, ELISA Standard
Formulation Protein with no preservative
Gene ID (Entrez) 10218
Molecular Weight (g/mol) 38.2 kDa
Name Human ANGPTL-7
Purification Method Purified
Quantity 500 μg
Source Insect cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias angiopoietin like 7; angiopoietin-like 7; Angiopoietin-like factor; angiopoietin-like factor (CDT6); angiopoietin-like protein 7; Angiopoietin-related protein 7; ANGPTL7; AngX; CDT6; Cornea-derived transcript 6 protein; dJ647M16.1; UNQ313/PRO356
Common Name ANGPTL7
Gene Symbol ANGPTL7
Biological Activity Measured by its binding ability to recombinant alphavbeta3 integrin in a functional ELISA.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag
Sequence QKLSKHKTPA QPQLKAANCC EEVKELKAQV ANLSSLLSEL NKKQERDWVS VVMQVMELES NSKRMESRLT DAESKYSEMN NQIDIMQLQA AQTVTQTSAD AIYDCSSLYQ KNYRISGVYK LPPDDFLGSP ELEVFCDMET SGGGWTIIQR RKSGLVSFYR DWKQYKQGFG SIRGDFWLGN EHIHRLSRQP TRLRVEMEDW EGNLRYAEYS HFVLGNELNS YRLFLGNYTG NVGNDALQYH NNTAFSTKDK DNDNCLDKCA QLRKGGYWYN CCTDSNLNGV YYRLGEHNKH LDGITWYGWH GSTYSLKRVE MKIRPEDFKP HHHHHHHH
Content And Storage -20°C
Activity Measured by its binding ability to recombinant alphavbeta3 integrin in a functional ELISA.
Expression System Insect cells
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.