missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Ankyrin B (aa 3571-3706) Control Fragment Recombinant Protein

Artikelnummer. 30199916
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μL
missing translation for 'unitSize'
100µL
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 30199916

missing translation for 'mfr': Invitrogen™ RP102423

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82326 (PA5-82326. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the ankyrin family of proteins that link the integral membrane proteins to the underlying spectrin-actin cytoskeleton. Ankyrins play key roles in activities such as cell motility, activation, proliferation, contact and the maintenance of specialized membrane domains. Most ankyrins are typically composed of three structural domains: an amino-terminal domain containing multiple ankyrin repeats; a central region with a highly conserved spectrin binding domain; and a carboxy-terminal regulatory domain which is the least conserved and subject to variation. The protein encoded by this gene is required for targeting and stability of Na/Ca exchanger 1 in cardiomyocytes. Mutations in this gene cause long QT syndrome 4. Multiple transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Spezifikation

Accession Number Q01484
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 287
Name Human Ankyrin B (aa 3571-3706) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI835472; Ank2; ANK-2; Ank3; AnkB; ankyrin 2, brain; ankyrin 2, neuronal; ankyrin 3, epithelial; ankyrin B; ankyrin, brain; ankyrin-2; ankyrin-2, nonerythrocytic; ankyrin-B; AW491075; Brain ankyrin; brank-2; LQT4; Non-erythroid ankyrin
Common Name Ankyrin B
Gene Symbol ANK2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ERLAYIADHLGFSWTELARELDFTEEQIHQIRIENPNSLQDQSHALLKYWLERDGKHATDTNLVECLTKINRMDIVHLMETNTEPLQERISHSYAEIEQTITLDHSEGFSVLQEELCTAQHKQKEEQAVSKESETC
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt