missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human Apo-SAA1 Recombinant Protein, PeproTech®
Beskrivning
300-53-1MG will be provided as 2 x 500 μg (300-53-500UG). Recombinant Human Apo-SAA1 is an 11.7 kDa protein containing 105 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis.
The serum amyloid A family comprises a number of differentially expressed apolipoproteins, acute-phase SAA1 and SAA2, the former being the major component in plasma and constitutive SAAs. Although the liver is the primary site of synthesis of both SAA types extrahepatic production has been reported. The in vivo concentrations increase by as much as 1000 fold during inflammation. Several studies have expressed its importance in the diagnosis and monitoring of various diseases. Pathological SAA values are often detected in association with normal CRP concentrations; SAA rises earlier and more sharply than CRP. Recently, a broader view of SAA expression and function has been emerging.
Specifikationer
Specifikationer
| Accession Number | P0DJI8 |
| For Use With (Application) | Control, Bioactivity |
| Format | Lyophilized |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 6288 |
| Molecular Weight (g/mol) | 11.7 kDa |
| Name | Human Apo-SAA1 |
| Purification Method | Purified |
| Quantity | 2 x 500 μg |
| Storage Requirements | -20°C |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?