missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ARL17A (aa 74-122) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (27%), Rat (27%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61339 (PA5-61339. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
GTP-binding protein that functions as an allosteric activator of the cholera toxin catalytic subunit, an ADP-ribosyltransferase. Involved in protein trafficking; may modulate vesicle budding and uncoating within the Golgi apparatus. [UniProt]
Specifications
Specifications
| Accession Number | Q8IVW1 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 51326 |
| Name | Human ARL17A (aa 74-122) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | ADP ribosylation factor like GTPase 17 A; ADP-ribosylation factor 7; ADP-ribosylation factor 7 variant; ADP-ribosylation factor-like 17-like; ADP-ribosylation factor-like protein 17; ARF1P2; ARF1P2 ARL17A; ARL17A; ARL17B; ARL17P1; PRO2667 |
| Common Name | ARL17A |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction