missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human B3GALNT1 (NP_003772) VersaClone cDNA

Product Code. 18700808
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18700808 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18700808 Supplier R&D Systems™ Supplier No. RDC2582
  • ONLINE EXCLUSIVE PRICE
    328.00 €
    309.75 € / 10µg
    Save 18.25 €
    5% Off
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.

  • B3GALNT1 beta-1,3-Nacetylgalactosaminyltransferase 1 (globoside blood group) [ Homo sapiens (human) ]
  • Also known as: P; P1; GLOB; GLCT3; galT3; Gb4Cer; B3GALT3; beta3Gal-T3
  • B3GALNT1 is a member of the beta1,3-galactosyltransferase (beta3GalT) gene family.
  • This family encodes type II membrane-bound glycoproteins with diverse enzymatic functions using different donor substrates (UDP-galactose and UDP-N-acetylglucosamine) and different acceptor sugars (Nacetylglucosamine, galactose, Nacetylgalactosamine).
  • B3GALNT1 does not use N-acetylglucosamine as an acceptor sugar at all.
  •  The B3GALNT1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
  • For reasearch use only. Not for use in humans.
TRUSTED_SUSTAINABILITY

Specifications

Gene B3GALNT1
Accession Number NP_003772
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Insert Type 1009 bp
Gene ID (Entrez) 8706
Gene Alias b3Gal-T3, B3GALT3, Beta-1,3-galactosyltransferase 3, Beta-1,3-GalNAc-T1, Beta-1,3-GalTase 3, beta-1,3-N-acetylgalactosaminyltransferase 1 (globoside blood group), Beta3GalT3, Beta3Gal-T3, beta-3-Gx-T3, brainiac1, EC 2.4.1.79, Galactosylgalactosylglucosylceramide beta-D-acetyl-galactosaminyltransferase, Gb4Cer, GLCT3, Globoside Synthase, globotriaosylceramide 3-beta-N-acetylgalactosaminyltransferase, P, P antigen synthase, P blood group globoside, polypeptide 3 (Globosideblood group), UDP-GalNAc:beta-1,3-N-acetylgalactosaminyltransferase 1, UDP-GalNAc:betaGlcNAc beta 1,3-galactosaminyltransferase, polypeptide 1(Globoside blood group), UDP-GalNAc:betaGlcNAc beta-1,3-galactosaminyltransferase, polypeptide 1(Globoside blood group), UDP-N-acetylgalactosamine:globotriaosylceramidebeta-1,3-N-acetylgalactosaminyltransferase
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.