missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human BAFF/BLyS/TNFSF13B Quantikine ELISA Kit

Product Code. p-4753522
Click to view available options
Quantity:
1 Plate
6 Plates
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18761974

Brand: R&D Systems™ DBLYS0B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human BAFF/BLyS/TNFSF13B Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human BAFF/BLyS/TNFSF13B Quantikine ELISA Kit quantifies human BAFF/BLyS/TNFSF13B in cell culture supernates (25 ul), serum (25 ul), edta plasma (25 ul), heparin plasma (25 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 62.50 - 4000pg/mL
Assay Sensitivity 6.44pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Specificity Natural and Recombinant Human BAFF/BLyS
Sufficient For 1 x 96-well Plate
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 10673
Quantity 1 Plate
Sample Volume Cell Culture Supernates (25uL),Serum (25uL),EDTA Plasma (25uL),Heparin Plasma (25uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym ApoL related ligand TALL-1, B lymphocyte stimulator, BAFFB-cell activating factor, B-cell-activating factor, BLyS, BLYSB-lymphocyte stimulator, CD257, CD257 antigen, Dendritic cell-derived TNF-like molecule, DTL, TALL1, TALL-1 delta BAFF, TALL1Delta4 BAFF, THANK, TNF and ApoL-related leukocyte expressed ligand 1, TNF- and APOL-related leukocyte expressed ligand 1, TNF homolog that activates apoptosis, TNFSF13B, TNFSF20, tumor necrosis factor (ligand) superfamily, member 13 b, tumor necrosis factor (ligand) superfamily, member 20, tumor necrosis factor ligand superfamily member 13 B, tumor necrosis factor superfamily, member 13 b, tumor necrosis factor-like protein ZTNF4, ZTNF4
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.