Learn More
R&D Systems™ Human beta-Arrestin 2 (NP_004304.1) VersaClone cDNA
Product Code. 18722037
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. Quantity Unit Size
18722037 10 μg 10µg
1 options
Product Code. 18722037 Supplier R&D Systems™ Supplier No. RDC3623
  • ONLINE EXCLUSIVE PRICE
    287.00 €
    270.90 € / 10µg
    Save 16.10 €
    5% Off
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Gene beta-Arrestin 2
Accession Number NP_004304.1
Gene ID (Entrez) 409
Gene Alias ARB2, ARR2, arrestin 3, Arrestin beta-2, arrestin, beta 2, BARR2, beta-arrestin-2, DKFZp686L0365
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.