missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Biomarker Panel A Luminex™ Performance Control

Product Code. 18461170
Click to view available options
Quantity:
1 Ea.
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18461170

Brand: R&D Systems™ QC07

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.

  • For use as quantitative controls for the determination of human cytokine concentrations in biological fluids.
  • Concentrations have been assigned using R&D Systems Luminex™ High Performance Assays.
  • Controls are prepared in a buffered protein solution with preservatives. They contain recombinant human cytokines at low, medium and high concentrations. Controls are supplied lyophilized.
TRUSTED_SUSTAINABILITY

Specifications

Product Type ELISA Kit
Quantity 1 Ea.
Species Human
Binding Target Luminex Assay Controls
For Use With (Equipment) Human Biomarker Panel A
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.