missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BOLL Partial ORF (NP_149019.1, 80 a.a. - 176 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00066037-Q03.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16045645

  • 508.00 € / 25µg
Estimated Shipment: 23-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene belongs to the DAZ gene family required for germ cell development. It encodes an RNA-binding protein which is more similar to Drosophila Boule than to human proteins encoded by genes DAZ (deleted in azoospermia) or DAZL (deleted in azoospermia-like). Loss of this gene function results in the absence of sperm in semen (azoospermia). Histological studies demonstrated that the primary defect is at the meiotic G2/M transition. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq]

Sequence: FETQEDAQKILQEAEKLNYKDKKLNIGPAIRKQQVGIPRSSIMPAAGTMYLTTSTGYPYTYHNGVAYFHTPEVTSVPPPWPSRSVCSSPVMVAQPIY
Specifications

Specifications

NP_149019.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.3kDa
12.5% SDS-PAGE Stained with Coomassie Blue
FETQEDAQKILQEAEKLNYKDKKLNIGPAIRKQQVGIPRSSIMPAAGTMYLTTSTGYPYTYHNGVAYFHTPEVTSVPPPWPSRSVCSSPVMVAQPIY
RUO
BOLL
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
66037
BOLL (Human) Recombinant Protein (Q03)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BOLL
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers