missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Brain Membrane Tissue Lysate (Adult Membrane Alzheimer's)

Product Code. 18280140
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18280140 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18280140 Supplier Novus Biologicals™ Supplier No. NBL126447
  • ONLINE EXCLUSIVE PRICE
    779.00 €
    736.05 € / 0.1mg
    Save 42.95 €
    5% Off
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
The Human Brain Membrane Tissue Lysate (Adult Membrane Alzheimer's) is derived from human.
TRUSTED_SUSTAINABILITY

Specifications

Tissue Brain
Quantity 0.1 mg
Storage Buffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Test Specificity Membrane Protein - Alzheimers Disease: Brain: Temporal Lobe
Cellular Fraction Membrane
Tissue Condition Alzheimer's
Cross Reactivity Human
Host Species Human
Product Type Human Brain Membrane Tissue Lysate (Adult Alzheimer's)
Content And Storage -80°C. Avoid freeze-thaw cycles.
Life Stage Adult
Protein State Native
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.