missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human BRSK1 Synthetic Peptide

Product Code. 15286679
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15286679 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15286679 Supplier Invitrogen™ Supplier No. PEP0472
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Synthetic Peptide

This peptide corresponds to 28 amino acids near the carboxy terminus of human BRSK1 PEP-0472 can be used as a blocking peptide with polyclonal antibody PA5-20352.

BRSK1 (SAD1) is a serine/threonine kinase, and is a UV-induced DNA damage checkpoint kinase. BRSK1 is unbiquitously expressed, with highest levels of expression in the brain and testes. Similar to its yeast homolog, BRSK1 is thought to be involved in stress-induced cell cycle arrest. Overexpression of this protein leads to the G2/M arrest in HeLa S2 cells and UV-induced G2/M arrest could be partially abrogated by reduced expression of BRSK1 through the use of siRNA, indicating its role in DNA damage checkpoint function. More recently, it has been shown that both BRSK1 and the related protein BRSK2 are required for mammalian neuronal polarization. While BRSK1- and BRSK2-null mice were viable, double-mutant mice died within two hours of birth. Neurons from these mice showed uniformly-sized neurites as opposed to the normal long axon and multiple shorter dendrites. These neurites also displayed both axonal and dendritic markers. At least two isoforms of BRSK1 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 84446
Species Human
Purification Method purified
Accession Number Q8TDC3
Common Name BRSK1
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias BR serine/threonine kinase 1; BR serine/threonine-protein kinase 1; Brain-selective kinase 1; brain-specific serine/threonine-protein kinase 1; Brsk1; Gm1100; hSAD1; KIAA1811; protein kinase SAD1A; RGD1563268; SAD1; SAD1 homolog; SAD1 kinase; SAD1A; SADB; SAD-B; SadB kinase short isoform; serine/threonine kinase SADB; serine/threonine kinase SAD-B; serine/threonine-protein kinase BRSK1; Serine/threonine-protein kinase SAD-B; synapses of Amphids Defective homolog 1
Gene Symbol Brsk1
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.