missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human BRWD1 (aa 1576-1670) Control Fragment Recombinant Protein

Product Code. 30199061
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30199061 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30199061 Supplier Invitrogen™ Supplier No. RP95905

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (82%), Rat (82%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56597 (PA5-56597. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

WDR9.3 encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This protein contains 2 bromodomains and multiple WD repeats, and the function of this protein is not known. This gene is located within the Down syndrome region 2 on chromosome 21. Alternative splicing of this gene generates 3 transcript variants diverging at the 3' ends.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NSI6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 54014
Name Human BRWD1 (aa 1576-1670) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 5330419I02Rik; bromodomain and WD repeat domain containing 1; bromodomain and WD repeat-containing protein 1; BRWD1; C21orf107; D530019K20Rik; DCAF19; FLJ43918; G1-403-16; N143; repro5; transcriptional unit N143; WD repeat domain 9; WD repeat protein WDR9-form2; WD repeat-containing protein 9; WDR9
Common Name BRWD1
Gene Symbol Brwd1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence RTRAAQRKTGPVSLANGCGRKATRKRVYLSDSDNNSLETGEILKARAGNNRKVLRKCAAVAANKIKLMSDVEENSSSESVCSGRKLPHRNASAVA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.