missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human C19orf61 (aa 329-413) Control Fragment Recombinant Protein

Product Code. 30181555
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30181555 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30181555 Supplier Invitrogen™ Supplier No. RP98993

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59714 (PA5-59714. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a T cell adaptor protein, a class of intracellular molecules with modular domains capable of recruiting additional proteins but that exhibit no intrinsic enzymatic activity. The encoded protein contains a unique N-terminal region followed by a PH domain and C-terminal SH3 domain. Along with the adhesion and degranulation-promoting adaptor protein, the encoded protein plays a critical role in inside-out signaling by coupling T-cell antigen receptor stimulation to the activation of integrins.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9H0W8
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 56006
Name Human C19orf61 (aa 329-413) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1500002O20Rik; AI429693; C19orf61; F17127_1; HBMS; N28092; protein SMG9; Protein SMG-9; protein smg-9 homolog; RGD1309888; Smg9; smg-9 homolog, nonsense mediated mRNA decay factor (C. elegans); SMG9 nonsense mediated mRNA decay factor; SMG9, nonsense mediated mRNA decay factor
Common Name C19orf61
Gene Symbol SMG9
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SLYRFLQTAEMVKPSTPSPSHESSSSSGSDEGTEYYPHLVFLQNKARREDFCPRKLRQMHLMIDQLMAHSHLRYKGTLSMLQCNV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.