missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Calreticulin (NP_004334.1) VersaClone cDNA

Product Code. 18734527
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18734527 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18734527 Supplier R&D Systems™ Supplier No. RDC3357
  • ONLINE EXCLUSIVE PRICE
    328.00 €
    309.75 € / 10µg
    Save 18.25 €
    5% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Contains gene inserts that are fully sequenced and produced with high quality

TRUSTED_SUSTAINABILITY

Specifications

Gene Calretinin
Accession Number NP_004334.1
Concentration 10 μg (at 0.2 μg/μL)
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Insert Type 1267 bp
Gene ID (Entrez) 794
Gene Alias 29 kDa calbindin, CAB29, CAL2, CALB2, calbindin 2, calbindin 2, (29 kD, calretinin), calbindin 2, 29 kDa (calretinin), calbindin D29K, calretinin, CR
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Product Line R&D Systems
Quantity 10 μg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.