missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Caspase 5 Synthetic Peptide

Product Code. 15226449
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15226449 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15226449 Supplier Invitrogen™ Supplier No. PEP0229
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Synthetic Peptide

Caspase-5 Synthetic Peptide, PEP-0229, from Invitrogen.

This peptide corresponds to 14 amino acids near the center of human Caspase-5.PEP-0229 can be used as a blocking peptide with polyclonal antibody PA5-20111.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 838
Species Human
Purification Method purified
Accession Number P51878
Common Name Human Caspase 5
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol CASP5
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.