Learn More
Abnova™ Human CAST Partial ORF (NP_001741.3, 582 a.a. - 669 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_001741.3 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 831 |
Molecular Weight (g/mol) | 35.42kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16043715
|
Abnova™
H00000831-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16033715
|
Abnova™
H00000831-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is an endogenous calpain (calcium-dependent cysteine protease) inhibitor. It consists of an N-terminal domain L and four repetitive calpain-inhibition domains (domains 1-4), and it is involved in the proteolysis of amyloid precursor protein. The calpain/calpastatin system is involved in numerous membrane fusion events, such as neural vesicle exocytosis and platelet and red-cell aggregation. The encoded protein is also thought to affect the expression levels of genes encoding structural or regulatory proteins. Several alternatively spliced transcript variants of this gene have been described, but the full-length natures of only some have been determined. [provided by RefSeq]
Sequence: QPDPDENKPMEDKVKEKAKAEHRDKLGERDDTIPPEYRHLLDDNGQDKPVKPPTKKSEDSKKPADDQDPIDALSGDLDSCPSTTETSQSpecifications
NP_001741.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.42kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BS-17/MGC9402 | |
CAST | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
831 | |
CAST (Human) Recombinant Protein (Q01) | |
QPDPDENKPMEDKVKEKAKAEHRDKLGERDDTIPPEYRHLLDDNGQDKPVKPPTKKSEDSKKPADDQDPIDALSGDLDSCPSTTETSQ | |
RUO | |
CAST | |
Wheat Germ (in vitro) | |
GST | |
Liquid |