missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CAT1 (aa 473-567) Control Fragment Recombinant Protein

Product Code. 30200423
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30200423

Brand: Invitrogen™ RP92890

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (91%), Rat (91%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-110860 (PA5-110860. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Carnitine acetyltransferase (CRAT) is a key enzyme in the metabolic pathway in mitochondria, peroxisomes and endoplasmic reticulum. CRAT catalyzes the reversible transfer of acyl groups from an acyl-CoA thioester to carnitine and regulates the ratio of acylCoA/CoA in the subcellular compartments. Different subcellular localizations of the CRAT mRNAs are thought to result from alternative splicing of the CRAT gene suggested by the divergent sequences in the 5' region of peroxisomal and mitochondrial CRAT cDNAs and the location of an intron where the sequences diverge. The alternatively splicing of this gene results in three distinct isoforms, one of which contains an N-terminial mitochondrial transit peptide, and has been shown to be located in mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P43155
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 1384
Name Human CAT1 (aa 473-567) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias amino acid transporter, cationic 1; ATRC1; AW107812; CARAT; Carnitine acetylase; carnitine acetyltransferase; Carnitine O-acetyltransferase; CAT; CAT1; CAT-1; Crat; Ecotropic retroviral leukemia receptor homolog; ecotropic retroviral receptor; ecotropic retrovirus receptor homolog; ERR; HCAT1; High affinity cationic amino acid transporter 1; REC1L; SLC7A1; solute carrier family 7 (cationic amino acid transporter, y+ system), member 1; solute carrier family 7 member 1; System Y+ basic amino acid transporter
Common Name CAT1
Gene Symbol CRAT
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MDSLTFVKAMDDSSVTEHQKVELLRKAVQAHRGYTDRAIRGEAFDRHLLGLKLQAIEDLVSMPDIFMDTSYAIAMHFHLSTSQVPAKTDCVMFFG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.