missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD19 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
191.00 € - 344.00 €
Specifications
| Antigen | CD19 |
|---|---|
| Clone | 4G7-2E3 |
| Dilution | Flow Cytometry 5 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
CD19 Monoclonal specifically detects CD19 in Human samples. It is validated for Flow Cytometry.Specifications
| CD19 | |
| Flow Cytometry 5 uL/10^6 cells | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 930 | |
| Human CLL cells | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| 4G7-2E3 | |
| Flow Cytometry | |
| Alexa Fluor 594 | |
| Mouse | |
| Human | |
| B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, Leu-12, MGC12802, T-cell surface antigen Leu-12 | |
| CD19 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human CD19. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title