Learn More
R&D Systems™ Human TMIGD2/CD28H (NP_653216) VersaClone cDNA
Transmembrane and immunoglobulin domain containing 2
Brand: R&D Systems™ RDC0929
Description
The Human TMIGD2/CD28H (NP_653216) VersaClone cDNA clone contains the complete ORF for TMIGD2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The TMIGD2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
| TMIGD2 | |
| 126259 | |
| Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| NP_653216 | |
| CD28H, IGPR1, IGPR-1, MGC23244, transmembrane and immunoglobulin domain containing 2, transmembrane and immunoglobulin domain-containing protein 2 | |
| Human |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Reasearch Use Only; Not for Use in Humans