missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CD3 epsilon Fc Chimera Recombinant Protein
Click to view available options
Quantity:
50 μg
500 μg
Unit Size:
500µg
50µg
Description
It is recommended that the protein be aliquoted for optimal storage. Reconstituted with sterile deionized water to 0.25mg/mL. Concentration is measured by UV.
The CD3 complex, composed of gamma, delta, epsilon, and zeta subunits, is essential for the assembly, trafficking, and surface expression of the T cell receptor (TCR) complex. These subunits are structurally related members of the immunoglobulin superfamily and are encoded by closely linked genes on human chromosome 11. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells, but not on B or NK cells. The CD3 subunits play a crucial role in transducing antigen-recognition signals into the cytoplasm of T cells. The cytoplasmic tails of CD3 subunits contain a double tyrosine-based motif that associates with cytoplasmic signal transduction molecules, mediating T cell activation through the TCR. Crosslinking of the TCR initiates intracellular biochemical pathways that result in cellular activation, proliferation, and potentially growth arrest and cell survival. CD3 is present on 68-82% of normal peripheral blood lymphocytes, 65-85% of thymocytes, and Purkinje cells in the cerebellum. Decreased percentages of T lymphocytes may be observed in some autoimmune diseases. Defects in the CD3 gene are associated with CD3 immunodeficiency, highlighting its importance in immune function and regulation.
Specifications
Specifications
| Accession Number | P04234, P07766, P09693, P20963 |
| Concentration | 1.43 mg/mL |
| For Use With (Application) | Control, Neutralization |
| Format | Lyophilized |
| Formulation | PBS with 5% mannitol, 5% trehalose, 0.01% Tween 80 and no preservative; pH 7.4 |
| Gene ID (Entrez) | 915, 916, 917, 919 |
| Molecular Weight (g/mol) | 38.5 kDa |
| Name | Human CD3 epsilon Fc Chimera |
| pH Range | 7.4 |
| Purification Method | Purified |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction