Learn More
Invitrogen™ Human CDCA3 (aa 152-252) Control Fragment Recombinant Protein
Beskrivning
Highest antigen sequence indentity to the following orthologs: Mouse (78%), Rat (78%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-110918 (PA5-110918. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifikationer
Specifikationer
| Accession Number | Q99618 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 83461 |
| Name | Human CDCA3 (aa 152-252) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 2410005A12Rik; C8; CDCA 3; CDCA3; cell division cycle associated 3; cell division cycle-associated protein 3; gene rich cluster, C8; gene-rich cluster protein C8; GRCC 8; GRCC8; MGC2577; TOME 1; Tome1; TOME-1; trigger of mitotic entry 1; trigger of mitotic entry protein 1 |
| Common Name | CDCA3 |
| Visa mer |
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.